Bromophenol blue 0.04% in water

Supplier: TCI America

Synonyms: Bromphenol Blue Sultone Form, 3,3',5,5'-Tetrabromophenolsulfonphthalein, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein, Tetrabromophenol-sulfonephthalein

B0117-500ML
TCB0117-500MLEA 57.02 USD
TCB0117-500ML
Bromophenol blue 0.04% in water
Bromophenol blue

[for pH Determination]
MDL Number: MFCD00005875

Formula: C₁₉H₁₀Br₄O₅S
MW: 669.97 g/mol
MDL Number: MFCD00005875
CAS Number: 115-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR