Bromocresol green 0.04% in ethanol 50%

Supplier: TCI America
Danger

B0114-500ML
TCB0114-500MLEA 75 USD
TCB0114-500ML
Bromocresol green 0.04% in ethanol 50%
Bromocresol green

[for Titration]
CAS Number: 76-60-8
MDL Number: MFCD00005874
Molecular Weight: 698.01
Form: Clear Liquid
Color: Yellow
Flash Point (°C): 24

Formula: C₂₁H₁₄Br₄O₅S
Flash Pt: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00005874
CAS Number: 76-60-8
EINECS: 200-972-8
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=318f12e011cacdefca0126ce40eeaf9d84d26ee9&vid=da4d68efa2de518b40e7c1e446a818b7eda5692f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR