Benzoxazole ≥98.0% (by GC)
Supplier: TCI America
B0095-25G
TCB0095-25GEA
77
USD
TCB0095-25G
Benzoxazole ≥98.0% (by GC)
Benzoxazole
CAS Number: 273-53-0
MDL Number: MFCD00005765
Molecular Formula: C7H5NO
Molecular Weight: 119.12
Purity/Analysis Method:
Formula:
C₇H₅NO Boiling Pt: 183 °C Flash Pt: 58 °C |
MDL Number:
MFCD00005765 CAS Number: 273-53-0 UN: 1325 ADR: 4.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...