Benzoic acid ≥99.0%, zone refined: number of passes 20

Supplier: TCI America
Warning

B0062-1SAMPLE
TCB0062-ZONEEA 86.8 USD
TCB0062-ZONE
Benzoic acid ≥99.0%, zone refined: number of passes 20
Benzoic acid

CAS Number: 65-85-0
MDL Number: MFCD00002398
Molecular Formula: C7H6O2
Molecular Weight: 122.12
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 249
Melting point (°C): 125
Flash Point (°C): 118

Formula: C₇H₆O₂
Boiling Pt: 249 °C
Melting Pt: 125 °C
Density: 1.316 g/cm³ (20 °C)
Flash Pt: 118 °C
Storage Temperature: Ambient
MDL Number: MFCD00002398
CAS Number: 65-85-0
EINECS: 200-618-2
REACH: 01-2119455536-33
Merck Index: 13,01092
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43c224bb45b8b5722e9440025ae967a22ebc99c5&vid=f9f2baa8a39e51cc559bc22a6cf0c9ada51c0cb9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR