Benzaldehyde azine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
CAS Number: 588-68-1
MDL Number: MFCD00016876
Molecular Formula: C14H12N2
Molecular Weight: 208.26
Purity/Analysis Method:
Other Product Names / Synonyms: 1-Benzaldehyde Azine
Formula:
C₁₄H₁₂N₂ Melting Pt: 93 °C |
MDL Number:
MFCD00016876 CAS Number: 588-68-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...