Dipropylammonium acetate ∼0.5 mol/L in water

Supplier: TCI America

Synonyms: IPC-DPAA

A5703-10ML
TCA5703-10MLEA 33 USD
TCA5703-10ML
Dipropylammonium acetate ∼0.5 mol/L in water
Dipropylammonium acetate

[Ion-Pair Reagent for LC-MS]
CAS Number: 114389-69-4
MDL Number: MFCD03093617
Molecular Formula: C8H19NO2
Molecular Weight: 161.25
Form: Clear Liquid
Color: Colorless


Other Product Names / Synonyms: IPC-DPAA

Formula: C₈H₁₉NO₂
MW: 161.25 g/mol
MDL Number: MFCD03093617
CAS Number: 114389-69-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd46578514007e4ee519c7958ede5fa726228581&vid=8ac34cef2f956495dba84a63035436596764cdd7&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR