N,O-Bis(trimethylsilyl)acetamide 25% in acetonitrile (TMS-BA) for gas chromatography

Supplier: TCI America

Synonyms: BSA, Silyl Derivatisation, Trimethylsilyl N-(trimethylsilyl)acetimidate, Trimethylsilyl N-trimethylsilylacetamidate

A5602-5ML
TCA5602-5MLEA 64.9 USD
TCA5602-5ML
N,O-Bis(trimethylsilyl)acetamide 25% in acetonitrile (TMS-BA) for gas chromatography
N,O-Bis(trimethylsilyl)acetamide

[for Gas Chromatography]
CAS Number: 10416-59-8
MDL Number: MFCD00008270
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 10
Storage Temperature: 0-10°C


Other Product Names / Synonyms: N,O-Bis(trimethylsilyl)acetamide, BSA

Formula: C₈H₂₁NOSi₂
MW: 203.43 g/mol
Flash Pt: 10 °C
MDL Number: MFCD00008270
CAS Number: 10416-59-8
UN: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4af01cc988d3dffedff20efec6749835637a9ae7&vid=488b47cbb2d623e540081d7db9f2a29ff58fbf8c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR