N,O-Bis(trimethylsilyl)acetamide ≥80.0% (by GC) for gas chromatography

Supplier: TCI America

Synonyms: BSA, Silyl Derivatisation, Trimethylsilyl N-(trimethylsilyl)acetimidate, Trimethylsilyl N-trimethylsilylacetamidate

A5601-5ML
TCA5601-5MLEA 31 USD
TCA5601-5ML
N,O-Bis(trimethylsilyl)acetamide ≥80.0% (by GC) for gas chromatography
N,O-Bis(trimethylsilyl)acetamide

Form: Clear Liquid
Boiling point (°C): 73
Melting point (°C): -24
Flash Point (°C): 42


Product Categories: Analytical Science, Silylation (GC Derivatizing Reagents)
Other Product Names / Synonyms: N,O-Bis(trimethylsilyl)acetamide

Formula: C₈H₂₁NOSi₂
MW: 203.43 g/mol
Boiling Pt: 73 °C
Melting Pt: –24 °C
Flash Pt: 42 °C
MDL Number: MFCD00008270
CAS Number: 10416-59-8
UN: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4bf925a64e01ad166b861fc1357b7fe586d438f7&vid=a3fb4576c17540886dfc46863f891d4d7dd62a83&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR