DAABD-Cl [=4-[2-(Dimethylamino)ethylaminosulfonyl]-7-chloro-2,1,3-benzoxadiazole]
Supplier: TCI America
Synonyms:
7-Chloro-N-(2-(dimethylamino)ethyl)benzo[c][1,2,5]oxadiazole-4-sulfonamide, 4-[2-(Dimethylamino)ethylaminosulfonyl]-7-chloro-2,1,3-benzoxadiazole, 7-Chloro-N-(2-(dimethylamino)ethyl)benzo[c][1,2,5]oxadiazole-4-sulphonamide, 4-[2-(Dimethylamino)ethylaminosulphonyl]-7-chloro-2,1,3-benzoxadiazole
[for Proteome Analysis]
CAS Number: 664985-43-7
Molecular Formula: C10H13ClN4O3S
Molecular Weight: 304.75
Form: Crystal
Color: Pale Yellow
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 4-[2-(Dimethylamino)ethylaminosulfonyl]-7-chloro-2,1,3-benzoxadiazole
Formula:
C₁₀H₁₃ClN₄O₃S MW: 304.75 g/mol |
MDL Number:
MFCD08276273 CAS Number: 664985-43-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...