4-Chloro-7-nitrobenzofurazan ≥98.0% (by titrimetric analysis)
Supplier: TCI America
[for HPLC Labeling]
CAS Number: 10199-89-0
MDL Number: MFCD00005808
Molecular Formula: C6H2ClN3O3
Molecular Weight: 199.55
Purity/Analysis Method:
Product Categories: Analytical Science, Fluorescence Detection (HPLC Labeling Reagents), Fluorescence Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: 4-Chloro-7-nitrobenzofurazan, 4-Chloro-7-nitro-2,1,3-benzoxadiazole
Formula:
C₆H₂ClN₃O₃ Melting Pt: 98 °C |
MDL Number:
MFCD00005808 CAS Number: 10199-89-0 EINECS: 233-496-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...