(R)-(-)-NBD-Py-NCS [=(R)-(-)-4-(3-Isothiocyanatopyrrolidin-1-yl)-7-nitro-2,1,3-benzoxadiazole] ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
(R)-(-)-4-(3-Isothiocyanatopyrrolidin-1-yl)-7-nitro-2,1,3-benzoxadiazole, (R)-(-)-4-(3-Isothiocyanatopyrrolidin-1-yl)-7-nitrobenzofurazan
[HPLC Labeling Reagent for e.e. Determination]
CAS Number: 163927-29-5
MDL Number: MFCD02093434
Molecular Formula: C11H9N5O3S
Molecular Weight: 291.29
Purity/Analysis Method:
Product Categories: Analytical Science, e.e. Determination (HPLC Labeling Reagents), Fluorescence Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: (R)-(-)-4-(3-Isothiocyanatopyrrolidin-1-yl)-7-nitrobenzofurazan, (R)-(-)-4-(3-Isothiocyanatopyrrolidin-1-yl)-7-nitro-2,1,3-benzoxadiazole
Formula:
C₁₁H₉N₅O₃S MW: 291.29 g/mol |
MDL Number:
MFCD02093434 CAS Number: 163927-29-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...