(S)-(-)-NBD-Pro-COCl [=(S)-(-)-4-Nitro-7-(2-chloroformylpyrrolidin-1-yl)-2,1,3-benzoxadiazole]
Supplier: TCI America
[HPLC Labeling Reagent for e.e. Determination]
CAS Number: 159717-68-7
MDL Number: MFCD00191512
Molecular Formula: C11H9ClN4O4
Molecular Weight: 296.67
Form: Crystal
Color: Yellow
Storage Temperature: 0-10°C
Other Product Names / Synonyms: (S)-(-)-4-Nitro-7-(2-chloroformylpyrrolidin-1-yl)benzofurazan, (S)-(-)-4-Nitro-7-(2-chloroformylpyrrolidin-1-yl)-2,1,3-benzoxadiazole
Formula:
C₁₁H₉ClN₄O₄ |
MDL Number:
MFCD00191512 CAS Number: 159717-68-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...