(R)-(-)-NBD-APy[=(R)-(-)-4-Nitro-7-(3-aminopyrrolidin-1-yl)-2,1,3-benzoxadiazole] ≥98.0% (by HPLC)
Supplier: TCI America
[HPLC Labeling Reagent for e.e. Determination]
CAS Number: 143112-51-0
MDL Number: MFCD00191510
Molecular Formula: C10H11N5O3
Molecular Weight: 249.23
Purity/Analysis Method:
Product Categories: Analytical Science, e.e. Determination (HPLC Labeling Reagents), Fluorescence Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: (R)-(-)-4-Nitro-7-(3-aminopyrrolidin-1-yl)benzofurazan, (R)-(-)-4-Nitro-7-(3-aminopyrrolidin-1-yl)-2,1,3-benzoxadiazole
Formula:
C₁₀H₁₁N₅O₃ |
MDL Number:
MFCD00191510 CAS Number: 143112-51-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...