(S)-(+)-NBD-APy [=(S)-(+)-4-Nitro-7-(3-aminopyrrolidin-1-yl)-2,1,3-benzoxadiazole]
Supplier: TCI America
[HPLC Labeling Reagent for e.e. Determination]
CAS Number: 143112-52-1
MDL Number: MFCD00191509
Molecular Formula: C10H11N5O3
Molecular Weight: 249.23
Form: Crystal
Color: Yellow
Melting point (°C): 175
Specific rotation [a]20/D: 14.5 deg (C=0.2, CH3CN)
Other Product Names / Synonyms: (S)-(+)-4-Nitro-7-(3-aminopyrrolidin-1-yl)benzofurazan, (S)-(+)-4-Nitro-7-(3-aminopyrrolidin-1-yl)-2,1,3-benzoxadiazole
Formula:
C₁₀H₁₁N₅O₃ Melting Pt: 175 °C |
MDL Number:
MFCD00191509 CAS Number: 143112-52-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...