(R)-(-)-DBD-APy [=(R)-(-)-4-(N,N-Dimethylaminosulfonyl)-7-(3-aminopyrrolidin-1-yl)-2,1,3-benzoxadiazole] ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
(R)-(-)-4-(N,N-Dimethylaminosulfonyl)-7-(3-aminopyrrolidin-1-yl)benzofurazan, (R)-(-)-4-(N,N-Dimethylaminosulphonyl)-7-(3-aminopyrrolidin-1-yl)benzofurazan, (R)-(-)-4-(N,N-Dimethylaminosulfonyl)-7-(3-aminopyrrolidin-1-yl)-2,1,3-benzoxadiazole, (R)-(-)-4-(N,N-Dimethylaminosulphonyl)-7-(3-aminopyrrolidin-1-yl)-2,1,3-benzoxadiazole
[HPLC Labeling Reagent for e.e. Determination]
CAS Number: 143112-49-6
MDL Number: MFCD00191379
Molecular Formula: C12H17N5O3S
Molecular Weight: 311.36
Purity/Analysis Method:
Product Categories: Analytical Science, e.e. Determination (HPLC Labeling Reagents), Fluorescence Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: (R)-(-)-4-(N,N-Dimethylaminosulfonyl)-7-(3-aminopyrrolidin-1-yl)benzofurazan, (R)-(-)-4-(N,N-Dimethylaminosulfonyl)-7-(3-aminopyrrolidin-1-yl)-2,1,3-benzoxadiazole
Formula:
C₁₂H₁₇N₅O₃S MW: 311.36 g/mol Melting Pt: 130 °C |
MDL Number:
MFCD00191379 CAS Number: 143112-49-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...