(S)-(+)-DBD-APy [=(S)-(+)-4-(N,N-Dimethylaminosulfonyl)-7-(3-aminopyrrolidin-1-yl)-2,1,3-benzoxadiazole] ≥98.0% (by HPLC)
Supplier: TCI America
[HPLC Labeling Reagent for e.e. Determination]
CAS Number: 143112-50-9
MDL Number: MFCD00191378
Molecular Formula: C12H17N5O3S
Molecular Weight: 311.36
Purity/Analysis Method:
Product Categories: Analytical Science, e.e. Determination (HPLC Labeling Reagents), Fluorescence Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: (S)-(+)-4-(N,N-Dimethylaminosulfonyl)-7-(3-aminopyrrolidin-1-yl)benzofurazan, (S)-(+)-4-(N,N-Dimethylaminosulfonyl)-7-(3-aminopyrrolidin-1-yl)-2,1,3-benzoxadiazole
Formula:
C₁₂H₁₇N₅O₃S Melting Pt: 130 °C |
MDL Number:
MFCD00191378 CAS Number: 143112-50-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...