Nɑ-(5-Fluoro-2,4-dinitrophenyl)-L-leucinamide ≥98.0% (by HPLC)
Supplier: TCI America
A5523-100MG
TCA5523-100MGEA
155.44
USD
TCA5523-100MG
Nɑ-(5-Fluoro-2,4-dinitrophenyl)-L-leucinamide ≥98.0% (by HPLC)
Nalpha-(5-Fluoro-2,4-dinitrophenyl)-L-leucinamide
[HPLC Labeling Reagent for e.e. Determination]
CAS Number: 178065-29-7
MDL Number: MFCD03844761
Molecular Formula: C12H15FN4O5
Molecular Weight: 314.27
Purity/Analysis Method:
Formula:
C₁₂H₁₅FN₄O₅ MW: 314.27 g/mol Melting Pt: 170 °C |
MDL Number:
MFCD03844761 CAS Number: 178065-29-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...