Sanger's reagent ≥99.0% (by GC)

Supplier: TCI America

Synonyms: 1,3-Dinitro-4-fluorobenzene, 2,4-Dinitrofluorobenzene, 1-Fluoro-2,4-dinitrobenzene

A5512-5G
TCA5512-5GEA 66.62 USD
TCA5512-5G
Sanger's reagent ≥99.0% (by GC)
Sanger's reagent

[for HPLC Labeling] CAS Number: 70-34-8 MDL Number: MFCD00007056 Molecular Formula: C6H3FN2O4 Molecular Weight: 186.10 Purity/Analysis Method: 99.0% (GC) Form: Crystal Boiling point (°C): 296 Melting point (°C): 26 Flash Point (°C): 164


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Analytical Science, UV Detection (HPLC Labeling Reagents) Other Product Names / Synonyms: 1-Fluoro-2,4-dinitrobenzene

Formula: C₆H₃FN₂O₄
MW: 186.1 g/mol
Boiling Pt: 296 °C
Melting Pt: 26 °C
Flash Pt: 164 °C
MDL Number: MFCD00007056
CAS Number: 70-34-8
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=476c9d60a2ebd87f0d3a78d425d6cd98c869bdba&vid=4ea8d766ead557265ec1f0d234325c4cc145810c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR