N-Chloromethyl-4-nitrophthalimide ≥98.0% (by titrimetric analysis)
Supplier: TCI America
[for HPLC Labeling]
CAS Number: 54455-34-4
MDL Number: MFCD00059889
Molecular Formula: C9H5ClN2O4
Molecular Weight: 240.60
Purity/Analysis Method:
Product Categories: Analytical Science, UV Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: 4-Nitro-N-chloromethylphthalimide
Formula:
C₉H₅ClN₂O₄ |
MDL Number:
MFCD00059889 CAS Number: 54455-34-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...