ω,4-Dibromoacetophenone ≥99.0%

Supplier: TCI America

Synonyms: Tau Protein Kinase I Inhibitor, 2,4'-Dibromoacetophenone, Glycogen Synthase Kinase-3β Inhibitor VII, α,4-Dibromoacetophenone, GSK-3β Inhibitor VII, TPK I Inhibitor, 2-Bromo-1-(4-bromophenyl)-1-ethanone, 4-Bromophenacyl bromide

A5501-5G
TCA5501-5GEA 116 USD
TCA5501-5G
ω,4-Dibromoacetophenone ≥99.0%
ω,4-Dibromoacetophenone

[for HPLC Labeling] CAS Number: 99-73-0 MDL Number: MFCD00000200 Molecular Formula: C8H6Br2O Molecular Weight: 277.94 Purity/Analysis Method: 99.0% (T) Form: Crystal Melting point (°C): 111 Storage Temperature: 0-10°C


Product Categories: Analytical Science, UV Detection (HPLC Labeling Reagents) Other Product Names / Synonyms: 2,4'-Dibromoacetophenone

Formula: C₈H₆Br₂O
MW: 277.94 g/mol
Boiling Pt: 310
Melting Pt: 111 °C
MDL Number: MFCD00000200
CAS Number: 99-73-0
UN: 3261
ADR: 8 II
Merck Index: 13,01412
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=437dca83c4051e8001fa2abf36ead32c7a23938c&vid=b1f3ae13de8e0c8eed83b4630868196b358c2501&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR