ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulfonate]) ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulfonic acid) diammonium salt, Ammonium 2,2'-(hydrazine-1,2-diylidene)bis(3-ethyl-2,3-dihydrobenzo[d]thiazole-6-sulfonate), ABTS, Ammonium 2,2'-(hydrazine-1,2-diylidene)bis(3-ethyl-2,3-dihydrobenzo[d]thiazole-6-sulphonate), 2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulphonic acid) diammonium salt

A5200-1G
TCA5200-001GEA 101.35 USD
TCA5200-001G
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulfonate]) ≥98.0% (by HPLC, titration analysis)
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate])

[Spectrophotometric reagent for free chlorine and use as chromogenic substrate in enzyme-immunoassay] CAS Number: 30931-67-0 MDL Number: MFCD00010404 Molecular Formula: C18H24N6O6S4 Molecular Weight: 548.67 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal


Other Product Names / Synonyms: 2,2'-Azinobis(3-ethylbenzothiazoline-6-sulfonic Acid Ammonium Salt)

Formula: C₁₈H₂₄N₆O₆S₄
MW: 548.68 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00010404
CAS Number: 30931-67-0
EINECS: 250-396-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53b101a63787263b9a828b847d96638de9f616e1&vid=a2087d20f0d7cd86b2f726512045dc0a70d90547&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR