1-Cetylpyridinium chloride monohydrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
[Ion association reagent for extraction photometric analysis and sensitizer in photometric determination of metals]
CAS Number: 6004-24-6
MDL Number: MFCD00149977
Molecular Formula: C21H38ClN
Molecular Weight: 339.99
Purity/Analysis Method:
Product Categories: Materials Chemistry, Cationic Surfactants, Analytical Science, Ion Association Reagents
Other Product Names / Synonyms: Cetylpyridinium Chloride, Hexadecylpyridinium Chloride
Formula:
C₂₁H₃₈ClN·1H₂O Melting Pt: 84 °C |
MDL Number:
MFCD00149977 CAS Number: 6004-24-6 UN: 2811 ADR: 6.1,I |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...