Benzyldimethyltetradecylammonium chloride hydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

A5160-5G
TCA5160-005GEA 46.81 USD
TCA5160-005G
Benzyldimethyltetradecylammonium chloride hydrate ≥98.0% (by HPLC, titration analysis)
Benzyldimethyltetradecylammonium chloride hydrate

[Ion association reagent for extraction photometric analysis and sensitizer in photometric determination of metals]
CAS Number: 139-08-2
MDL Number: MFCD00011771
Molecular Formula: C23H42ClN
Molecular Weight: 368.05
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Flash Point (°C): 160


Product Categories: Materials Chemistry, Cationic Surfactants, Analytical Science, Ion Association Reagents
Other Product Names / Synonyms: Tetradecyldimethylbenzylammonium Chloride

Formula: C₂₃H₄₂ClN·H₂O
MW: 368.05 g/mol (anhy)
Flash Pt: 160 °C
MDL Number: MFCD00011771
CAS Number: 139-08-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=142cea45fee7a754ef2b840da7118d4e251d287e&vid=145d7ff51e90694622f785b1dee98fc83e8a4941&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR