Methylene blue ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Basic Blue 9, Methylthioninium chloride, Methylene blue chloride, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride, Swiss Blue

A5105-1G
TCA5105-001GEA 40.7 USD
TCA5105-001G
Methylene blue ≥98.0% (by titrimetric analysis)
Methylene blue

[Ion association reagent for spectrophotometric analysis] CAS Number: 61-73-4 MDL Number: MFCD00012111 Molecular Formula: C16H18ClN3S Molecular Weight: 319.85 Purity/Analysis Method: 98.0% (T) Form: Crystal Lambda max.: 663 nm (EtOH(50vol%))


Product Categories: Bioscience, Stains & Dyes, Analytical Science, Ion Association Reagents Other Product Names / Synonyms: Basic Blue 9

Formula: C₁₆H₁₈ClN₃S
MW: 319.86 g/mol
Melting Pt: 180 °C
Density: 1.757 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150008
CAS Number: 61-73-4
EINECS: 200-515-2
Merck Index: 12,06137
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3727a13bc2e773cc967b924077d93f420e2c8b8b&vid=2f056ffb64fbdfda736b654daba6a021ea7fc7cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR