D-(-)-Luciferin ≥98.0%
Supplier: TCI America
Synonyms:
D-Luciferin, (S)-2-(6-Hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylic acid
[Chemiluminescence Reagent]
CAS Number: 2591-17-5
MDL Number: MFCD00042929
Molecular Formula: C11H8N2O3S2
Molecular Weight: 280.32
Purity/Analysis Method:
Product Categories: Bioscience, Luciferase Substrate, Analytical Science, Firefly Luciferin (Chemiluminescence Reagents)
Other Product Names / Synonyms: (S)-2-(6-Hydroxy-2-benzothiazolyl)-2-thiazoline-4-carboxylic Acid
Formula:
C₁₁H₈N₂O₃S₂ MW: 280.3228 g/mol Melting Pt: 186 °C |
MDL Number:
MFCD00042929 CAS Number: 2591-17-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...