1-(2-Pyridylazo)-2-naphthol (PAN) ≥98.0%

Supplier: TCI America

Synonyms: PAN, 1-(2-Pyridylazo)-2-naphthol

A5000-1G A5000-5G
TCA5000-001GEA 29.3 USD
TCA5000-001G TCA5000-005G
1-(2-Pyridylazo)-2-naphthol (PAN) ≥98.0%
1-(2-Pyridylazo)-2-naphthol

[Metal indicator and spectrophotometric reagent for transition metals] CAS Number: 85-85-8 MDL Number: MFCD00004071 Molecular Formula: C15H11N3O Molecular Weight: 249.27 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Melting point (°C): 142


Product Categories: Analytical Science, Azo Compounds (Chelating Agents), Azo Compounds (Chelating Agents) Other Product Names / Synonyms: 1-(2-Pyridylazo)-2-naphthol

Formula: C₁₅H₁₁N₃O
Boiling Pt: 412 °C (1013 hPa)
Melting Pt: 142 °C
Storage Temperature: Ambient
MDL Number: MFCD00004071
CAS Number: 85-85-8
EINECS: 201-637-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e877d4a93d602cfdf9942e75a38e298604a0e9ea&vid=ab5d238a929a937c0c3911d6f79f82599e71f5a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR