Acetanilide ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: N-Phenylacetamide

A3272-500G
TCA3272-500GEA 56 USD
TCA3272-500G
Acetanilide ≥98.0% (by GC)
Acetanilide
Formula: C₈H₉NO
MW: 135.17 g/mol
Boiling Pt: 304°C
MDL Number: MFCD00008674
CAS Number: 103-84-4
EINECS: 203-150-7
Merck Index: 14,00050

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 113.0 to 117.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR