Alizarin complexone ≥70% (by HPLC)
Supplier: TCI America
A3227-1G
A3227-5G
TCA3227-1GEA
41
USD
TCA3227-1G
TCA3227-5G
Alizarin complexone ≥70% (by HPLC)
Alizarin complexone
Formula:
C₁₉H₁₅NO₈ Melting Pt: 180 °C |
MDL Number:
MFCD00001202 CAS Number: 3952-78-1 EINECS: 223-544-2 |
Specification Test Results
Appearance | Orange to Amber to Dark red powder to crystal |
Purity (HPLC) | min. 70.0 area% |
Purity (Neutralization titration) | min. 80.0 % (calcd.on dried substance) |
Drying loss | max. 10.0 % |
Sensitiveness | Colorimetric Test : Turn Blue purple with Fluoride ion |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...