Alizarin complexone ≥70% (by HPLC)

Supplier: TCI America

A3227-1G A3227-5G
TCA3227-1GEA 41 USD
TCA3227-1G TCA3227-5G
Alizarin complexone ≥70% (by HPLC)
Alizarin complexone
Formula: C₁₉H₁₅NO₈
Melting Pt: 180 °C
MDL Number: MFCD00001202
CAS Number: 3952-78-1
EINECS: 223-544-2

Order Now

Specification Test Results

Appearance Orange to Amber to Dark red powder to crystal
Purity (HPLC) min. 70.0 area%
Purity (Neutralization titration) min. 80.0 % (calcd.on dried substance)
Drying loss max. 10.0 %
Sensitiveness Colorimetric Test : Turn Blue purple with Fluoride ion


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR