4-Amino-2-fluorobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

A3164-25G A3164-5G
TCA3164-25GEA 366 USD
TCA3164-25G TCA3164-5G
4-Amino-2-fluorobenzoic acid ≥98.0% (by GC, titration analysis)
4-Amino-2-fluorobenzoic acid
Formula: C₇H₆FNO₂
MW: 155.13
MDL Number: MFCD01569397
CAS Number: 446-31-1

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR