4-Acetylbenzenesulfonamide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

A3136-1G
TCA3136-1GEA 146 USD
TCA3136-1G
4-Acetylbenzenesulfonamide ≥98.0% (by HPLC, total nitrogen)
4-Acetylbenzenesulfonamide
Formula: C₈H₉NO₃S
MDL Number: MFCD00792524
CAS Number: 1565-17-9
EINECS: 216-363-5

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 178.0 to 182.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR