Azidoacetic acid ≥97.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Triazidoacetic acid, 2-Azidoacetic acid

A3079-1G
TCA3079-1GEA 76.83 USD
TCA3079-1G
Azidoacetic acid ≥97.0% (by GC)
Azidoacetic acid
Formula: C₂H₃N₃O₂
Boiling Pt: 116 °C (12 mmHg) (lit.)
Melting Pt: 16 °C (lit.)
Storage Temperature: Refrigerator
MDL Number: MFCD01320907
CAS Number: 18523-48-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6b85c9321c14d506a81e8edf8eba2c3383634cf&vid=d6b85c9321c14d506a81e8edf8eba2c3383634cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) Min. 97.0%
Refractive Index n20/D 1.4700 - 1.4740


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR