3'-Amino-4'-fluoroacetophenone ≥98.0% (by GC)

Supplier: TCI America
Warning

A2950-1G
TCA2950-1GEA 161 USD
TCA2950-1G
3'-Amino-4'-fluoroacetophenone ≥98.0% (by GC)
3'-Amino-4'-fluoroacetophenone
Formula: C₈H₈FNO
MDL Number: MFCD00115400
CAS Number: 2002-82-6

Order Now

Specification Test Results

Appearance White to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 69.0 to 73.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR