3'-Amino-2'-hydroxybiphenyl-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

A2944-200MG
TCA2944-1GEA 251.32 USD
TCA2944-1G TCA2944-200MG
3'-Amino-2'-hydroxybiphenyl-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)
3'-Amino-2'-hydroxybiphenyl-3-carboxylic acid
Formula: C₁₃H₁₁NO₃
MDL Number: MFCD10001467
CAS Number: 376592-93-7

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR