4-Anilino-1,1':4',1''-terphenyl ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

A2903-1G
TCA2903-1GEA 212 USD
TCA2903-1G
4-Anilino-1,1':4',1''-terphenyl ≥98.0% (by HPLC, total nitrogen)
4-Anilino-1,1':4',1''-terphenyl
Formula: C₂₄H₁₉N
MDL Number: MFCD28963769
CAS Number: 897671-81-7

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 193.0 to 197.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR