4-Amino-3-iodobenzoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

A2873-1G
TCA2873-1GEA 130 USD
TCA2873-1G
4-Amino-3-iodobenzoic acid ≥98.0% (by titrimetric analysis)
4-Amino-3-iodobenzoic acid
Formula: C₇H₆INO₂
MW: 263.03 g/mol
MDL Number: MFCD01660550
CAS Number: 2122-63-6

Order Now

Specification Test Results

Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 206.0 to 210.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR