5-Amino-3-methylisoxazole ≥97.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
3-Methylisoxazol-5-ylamine, 3-Methylisoxazol-5-amine
A2867-1G
A2867-5G
TCA2867-1GEA
60
USD
TCA2867-1G
TCA2867-5G
5-Amino-3-methylisoxazole ≥97.0% (by titrimetric analysis)
5-Amino-3-methylisoxazole
Formula:
C₄H₆N₂O MW: 98.1 g/mol Boiling Pt: 194 Melting Pt: 84 |
MDL Number:
MFCD00003151 CAS Number: 14678-02-5 EINECS: 238-719-9 |
Specification Test Results
Purity (GC) | min. 95.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 82.0 to 86.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...