Anti-SLeX Mouse Monoclonal Antibody [clone: 2H5]

Supplier: TCI America
A2849-1VIAL
TCA2849-VIALEA 841.1 USD
TCA2849-VIAL
Anti-SLeX Mouse Monoclonal Antibody [clone: 2H5]
Antibodies
Purified mouse monoclonal antibody/IgM, κ-chain

Type: Primary
Antigen: SLeX
Clonality: Monoclonal
Clone: 2H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR