Anti-Gb3 Mouse Monoclonal Antibody (Biotin) [clone: BGR23]

Supplier: TCI America
A2822-1VIAL
TCA2822-VIALEA 785.7 USD
TCA2822-VIAL
Anti-Gb3 Mouse Monoclonal Antibody (Biotin) [clone: BGR23]
Antibodies
Biotinylated mouse monoclonal antibody/IgG2b, κ-chain

Type: Primary
Antigen: Gb3
Clonality: Monoclonal
Clone: BGR23
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR