3-Aminofluoranthene ≥96.0% (by GC, titration analysis)

Supplier: TCI America
Warning

A2789-1G A2789-5G
TCA2789-1GEA 40 USD
TCA2789-1G TCA2789-5G
3-Aminofluoranthene ≥96.0% (by GC, titration analysis)
3-Aminofluoranthene
Formula: C₁₆H₁₁N
MDL Number: MFCD00001185
CAS Number: 2693-46-1
EINECS: 220-263-7
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to Amber powder to crystal
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 117.0 to 121.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR