Anti-GT1b Mouse Monoclonal Antibody [clone: GMRS]

Supplier: TCI America
A2732-1VIAL
TCA2732-VIALEA 673.7 USD
TCA2732-VIAL
Anti-GT1b Mouse Monoclonal Antibody [clone: GMRS]
Antibodies
Purified mouse monoclonal antibody/IgM, κ-chain

Type: Primary
Antigen: GT1b
Clonality: Monoclonal
Clone: GMRS
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR