Anti-GalNAc-GD1a Mouse Monoclonal Antibody [clone: 2A3D2]

Supplier: TCI America
A2701-1VIAL
TCA2701-VIALEA 716 USD
TCA2701-VIAL
Anti-GalNAc-GD1a Mouse Monoclonal Antibody [clone: 2A3D2]
Antibodies
Purified mouse monoclonal antibody/IgM, κ-chain

Specificity: Ganglioside GalNAc-GD1a. This antibody recognizes terminal GalNAcβ1→4(NeuAcα2→3)Galβ-R structure, and reacts to GM2, GalNAc-sialyl-nLc4, and Cad-active glycoproteins.

Type: Primary
Antigen: GalNAc-GD1a
Clonality: Monoclonal
Clone: 2A3D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR