Anti-SLeA Mouse Monoclonal Antibody [clone: 1H4]

Supplier: TCI America
A2584-1VIAL
TCA2584-VIALEA 393.4 USD
TCA2584-VIAL
Anti-SLeA Mouse Monoclonal Antibody [clone: 1H4]
Antibodies
Purified mouse monoclonal antibody/IgG3, κ-chain

Type: Primary
Antigen: SLeA
Clonality: Monoclonal
Clone: 1H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR