Anti-GM3 Mouse Monoclonal Antibody [clone: GMR6]

Supplier: TCI America
A2582-1VIAL
TCA2582-VIALEA 763.7 USD
TCA2582-VIAL
Anti-GM3 Mouse Monoclonal Antibody [clone: GMR6]
Antibodies
Purified mouse monoclonal antibody/IgM, κ-chain

Type: Primary
Antigen: GM3
Clonality: Monoclonal
Clone: GMR6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR