Anti-Lex Mouse Monoclonal Antibody [clone: 73-30]

Supplier: TCI America
A2578-1VIAL
TCA2578-VIALEA 665.4 USD
TCA2578-VIAL
Anti-Lex Mouse Monoclonal Antibody [clone: 73-30]
Antibodies
Purified mouse monoclonal antibody/IgM, κ-chain

Type: Primary
Antigen: Lex
Clonality: Monoclonal
Clone: 73-30
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR