Anti-GM2(NeuAc) Mouse Monoclonal Antibody [clone: MK1-16]

Supplier: TCI America
A2576-1VIAL
TCA2576-VIALEA 718.79 USD
TCA2576-VIAL
Anti-GM2(NeuAc) Mouse Monoclonal Antibody [clone: MK1-16]
Antibodies
Purified mouse monoclonal antibody/IgM, κ-chain

Type: Primary
Antigen: GM2(NeuAc)
Clonality: Monoclonal
Clone: MK1-16
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR