1,9-Pyrazoloanthrone ≥95.0% (by HPLC)

Supplier: TCI America

Synonyms: JNK Inhibitor II, Anthra[1,9-cd]pyrazol-6(2H)-one, Dibenzo[cd,g]indazol-6(2H)-one, SP600125, SP 600125

A2548-25MG
TCA2548-25MGEA 144.4 USD
TCA2548-25MG
1,9-Pyrazoloanthrone ≥95.0% (by HPLC)
1,9-Pyrazoloanthrone
Formula: C₁₄H₈N₂O
MW: 220.231 g/mol
Melting Pt: 282°C
Storage Temperature: Refrigerator
MDL Number: MFCD00022289
CAS Number: 129-56-6
EINECS: 204-955-6

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Green powder to crystal
Purity (HPLC) min. 95.0 area%
Elemental analysis (Nitrogen) 12.1 to 13.3 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR