Biotin-PEG3-azide ≥95.0%
Supplier: TCI America
Synonyms:
N-[2-[2-[2-(2-Azidoethoxy)ethoxy]ethoxy]ethyl]biotinamide, Azido-PEG3-biotin conjugate
A2523-100MG
TCA2523-100MGEA
385.58
USD
TCA2523-100MG
Biotin-PEG3-azide ≥95.0%
Biotin-PEG3-azide
CAS Number: 875770-34-6
MDL Number: MFCD20134145
Molecular Formula: C18H32N6O5S
Molecular Weight: 444.55
Purity/Analysis Method:
Formula:
C₁₈H₃₂N₆O₅S Melting Pt: 110 °C |
MDL Number:
MFCD20134145 CAS Number: 875770-34-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...