Anti-SLeA Mouse Monoclonal Antibody [clone: 2D3]

Supplier: TCI America
A2509-1VIAL
TCA2509-VIALEA 871.4 USD
TCA2509-VIAL
Anti-SLeA Mouse Monoclonal Antibody [clone: 2D3]
Antibodies
Purified mouse monoclonal antibody/IgM, κ-chain

Type: Primary
Antigen: SLeA
Clonality: Monoclonal
Clone: 2D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR