Anti-GD1b Mouse Monoclonal Antibody [clone: GGR12]

Supplier: TCI America
A2508-1VIAL
TCA2508-VIALEA 1032 USD
TCA2508-VIAL
Anti-GD1b Mouse Monoclonal Antibody [clone: GGR12]
Antibodies
Mouse monoclonal antibody/IgG3, κ-chain

Type: Primary
Antigen: GD1b
Clonality: Monoclonal
Clone: GGR12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR